Learn More
UVP Filter Glass: Shortwave UV Grid Replacement
Lamps have grid design, which is a highly uniform 254nm UV source of high-intensity
$894.00 - $1211.00
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
UVP77000104
|
UVP
77000104 |
Shortwave UV Grid | Model UVGD-68 Mineralight display lamp |
Each for $1,211.00
|
|
||||
UVP77000202
|
UVP
77000202 |
Shortwave Grid | Model Mineralight™ R-52G high intensity UV grid lamp |
Each for $894.00
|
|
||||
Description
- Lamp features a high intensity 254nm UV grid
- UVP's special UVG shortwave filter reduces solarization, prolonging the filter life
- Lamp mounts securely in its transformer base or is removable for movement around your work area
- Lamp manufactured with rugged, scratch-resistant plastic
- A snap-on filter assembly can be easily attached to convert this lamp for fluorescent analysis
Bacteria destruction, Sterilization, Photochemistry/Phosphorescence, Lapidary, Fluorescent analysis
Specifications
1 Ea. |