Learn More
UVP Filter Assemblies for White Light Transilluminators
Replacement filters for TW-26 and TW-43 models white light transilluminators
$402.00 - $456.00
Catalog Number | Mfr. No. | Filtration Area (English) | Filtration Area (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Filtration Area (English) | Filtration Area (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
UVP38016104
|
UVP
38016104 |
8.26 x 10.23 in. | 21 x 26 cm | TW-26 white light transilluminator |
Each for $402.00
|
|
|||
UVP38016501
|
UVP
38016501 |
14.1 x 16.9 in. | 36 x 43 cm | TW-43 white light transilluminator |
Each for $456.00
|
|
|||
Specifications
Filter Assembly |