Learn More
Honeywell™ Uvex™ Stealth™ Safety Goggles with Toric Lens
Click to view available options
Clear toric lens; Gray body
Description
Uvextreme™ anti-fog coating and indirect ventilation system minimize fogging
- Low profile design and indirect venting system for chemical splash and impact protection
- Frame and lens are impact resistant and provide greater than 99.9% protection from harmful UV rays
- Uvextreme anti-fog coating and indirect ventilation system minimize fogging
- Toric lens provides superior optics and unobstructed peripheral vision
- Soft, elastomer body flexes and conforms to the face for a secure, comfortable, gap-free fit
- Pivoting, quick-adjust headband provides easy adjustment for a comfortable fit
- One of the easiest and most cost effective lens replacement systems on the market
- Lens replacement system allows for multiple lens tints and extends the life of the goggle
- Rx insert provides flexibility for workers who require vision correction
- Clear lens; ideal for most indoor work applications
- Orange/gray body
- Headband: fabric
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
Lens Coating | Anti-fog |
Lens Type | Toric |
Lens Tint | Clear |
Frame Color | Gray |