Learn More
Honeywell™ Uvex™ Stealth™ OTG Goggles: Replacement Parts and Accessories
Click to view available options
Lens tint: clear
Description
For use with Uvex Stealth OTG goggles (Cat. No. 18-813-533)
- Ideal for impact, dust/airborne particles, chemical splash, and optical radiation (UV and welding/heat radiation)
- Superior optics and panoramic field of vision enhanced with toric lens
- Pivoting clip and quick-adjusting wide headband
- Elastomer body conforms and contours for a secure fit—even over most prescription eyewear
- Replacement lenses easily snap on
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
Lens Type | dura-streme anti-fog/Scratch-resistant |
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Replacement lens |
For Use With (Application) | Ideal for most indoor work applications |