Learn More
Honeywell™ Uvex™ Climazone™ Goggles
Click to view available options
Clear ultradura ext. lens
Description
- Designed for cold and extreme weather conditions
- Interior lens with Uvextreme anti-fog coating
- Eexterior lens with Ultra-dura scratch resistant hardcoat
- Not for chemical splash
- Meets ANSI Z87.1-2010 (High Impact) and is tested by Honeywell to meet the requirements of the CSA Z94.3 standard
Specifications
Lens Coating | Scratch-resistant |
Lens Type | Ultradura |
Lens Tint | Clear |
Product Type | Laboratory Goggles |
For Use With (Application) | For cold/extreme weather |