Learn More
Honeywell™ Uvex™ Classic™ Impact Goggles, Facefoam Model
Click to view available options
Hood indirect vent with facefoam model
Specifications
Lens Type | Uvextreme Anti-fog |
Lens Tint | Clear |
Product Type | Impact Goggle |
For Use With (Equipment) | Fits over most Rx spectacles |
For Use With (Application) | For dust or mist |
Includes | Facefoam, indirect vent |