Learn More
Honeywell™ Uvex™ Entity Safety Goggles
Click to view available options
Frame color: Translucent Blue; Lens tint: Clear; Uvextra AF lens coating
Description
- Toric shaped lens
- Lightweight
- Compact design to accommodate disposable respirators and other PPE
- PVC body conforms to face and provides comfortable, gap-free fit
- Protects against impact, dust and chemical splash hazards
- Clear lens tint with Uvextra™ anti-fog lens coating
- Adjustable, latex-free neoprene headband
- Protects against impact, dust and chemical splash
Applications:
Indoor applications.
Certifications
ANSI Z87.1-2010, CSA Z94.3 standard.
Specifications
Lens Coating | Anti-fog |
Lens Type | Uvextra AF |
Lens Tint | Clear |
Product Type | Safety goggles |
Frame Color | Translucent Blue |