Learn More
Honeywell™ Uvex™ Ambient OTG Safety Eyewear
Click to view available options
Lens Tint:
IR Filter Shade 3.0
Description
- The Uvex™ Ambient provides over-the-glass coverage, offering your workers full protection while wearing their own prescription eyewear
- Available with a range of comfort features and in two sizes, the Uvex™ Ambient OTG offers all-day coverage for a variety of workers and a wide range of prescription eyewear styles
- This high performance model can improve your workers' productivity as well as lower your cost-of-ownership with Uvex's renowned tints and lens replacement system
- The Uvex™ Ambient™ OTG is lightweight, comfortable and a stylish alternative to prescription safety eyewear
- Exceptional peripheral vision and coverage in a lightweight, wraparound uni-lens design
- Economical and easy lens replacement allows multiple tints and extends the life of the eyewear
Compliance
Meet ANSI™ Z87.1-2003. Certified to CSA Z94.3 standards.
Specifications
Lens Type | Ultra Dura |
Certifications/Compliance | ANSI Z87.1-2003 and CSA Z94.3 standards |
Lens Tint | IR Filter Shade 3.0 |
Product Type | Safety Eyewear; Over-the-glasses |
For Use With (Application) | Torch brazing operations |
Frame Color | Blue |
Size | Medium |
Lens Material | Polycarbonate |