Learn More
Honeywell™ Uvex™ 880 Series Klip Lifts For Hard Hat Visors
Click to view available options
Lens Tint:
Shade 3
Shade 6
Product Type:
880 Series Klip Lift, Full front
880 Series Klip Lift, Half front
Description
Whether you work in steelmaking, welding and cutting facilities, foundries or glassmaking operations, you will appreciate the convenience of the flip-up feature. These lenses can be quickly flipped down for observing a melt or a weld, and flipped back up again when not needed. Movement through a full 180° arc is possible, allowing the filter lenses to be raised or moved completely out of your line of vision.
- Quickly flip down to enhance vision, then flip up when not needed
- Integral spring clip holds 880 Series securely without screws, fasteners or tools
- Available with half or full lenses in cobalt blue shades 3, 4 and 6
Specifications
Lens Tint | Shade 6 |
Product Type | 880 Series Klip Lift, Half front |
For Use With (Equipment) | Hardhat visors |