Learn More
Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes, Standard
Designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Biotix™ uTIP™ Non-Filter Pipette Tips for Universal Pipettes are manufactured with low retention X-Resin™ giving tighter CV values.
Supplier: Biotix™ 63300050
Description
- Fits all standard pipettes
- FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
- X-Resin drastically reduces sample retention resulting in tighter CV values
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
Certifications
ISO 9001 , FDA Class I
Specifications
200 μL | |
Sterile | |
Autoclavable | |
Rack | |
uTip |
Non-Filtered | |
Yellow | |
e-beam | |
Yes | |
Polypropylene |