Learn More
Health O Meter UPRIGHT SCALE W/O WHEELS
SDP
Supplier:  Health O Meter 402KL
Encompass_Preferred
402KL Mechanical Beam Scale has a 390lb/180kg capacity and weighs in ¼ lb/100g increments. Includes a height rod that measures 23 5/8” - 84” / 60. Platform size is 10 ½” (w) x 14” (d) x 3 ¼” (h). Industry leading 10 Year Limited Warranty.
Catalog No. NC9088248