Learn More
Metrohm USA UNITRODE COMB PH 6.0259.100
Supplier:  Metrohm USA 60259100
Encompass
Metrohm Unitrode glass electrode is a high performance in difficult samples and at high temperatures. Bridge electrolyte type: c(KCl) is 3 mol/L. Diaphragm: fixed ground-joint diaphragm. Electrode plug-in head: Metrohm plug-in head G. Electrode slope > 0.97.
Catalog No. NC0646078