Learn More
United Scientific™ Neoprene Filter Adapter Set
SDPSCI_ED
This Neoprene Filter Adapter Set provides an alternative to special size holes in rubber stoppers.
Supplier:  United Scientific™ FASN07
Catalog No. S13106
Edge
Add to Cart
Description
Description
  • Set of 7 adapter cones designed to fit Buchner/fritted glass funnels, porcelain filters without headpieces, and for reverse flow cleaning of filter crucibles.
  • Used to support funnels, Gooch crucibles, filter tubes, and other apparatus in filtration assemblies.
  • Used individually or nested with adjacent sizes.
  • Functional even if not tightly fitted around funnel stem.
Specifications
Specifications
Buchner and fritted glass funnels, porcelain filters without headpieces, and for reverse flow cleaning of filter crucibles
Filter Adapter
Rubber
Filter Adapter Set of 7
Product Suggestions
Product Suggestions