Learn More
Simport™ Scientific UNISETTE™ Tissue Processing/Embedding Cassette
Secure specimens during processing with UNISETTE™ Tissue Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
$420.00 - $432.00
Specifications
Writing Face Angle | 35° |
---|---|
Material | Acetal |
Product Type | Tissue Processing/Embedding Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-045-011
|
Simport™ Scientific
M5052 |
White |
Case of 1500 for $432.00
|
|
|||||
22-045-012
|
Simport™ Scientific
M5053 |
Pink |
Case of 1500 for $432.00
|
|
|||||
22-045-013
|
Simport™ Scientific
M5054 |
Green |
Case of 1500 for $432.00
|
|
|||||
22-045-014
|
Simport™ Scientific
M5055 |
Yellow |
Case of 1500 for $432.00
|
|
|||||
22-045-015
|
Simport™ Scientific
M5056 |
Blue |
Case of 1500 for $420.00
|
|
|||||
22-045-016
|
Simport™ Scientific
M5057 |
Peach |
Case of 1500 for $432.00
|
|
|||||
22-045-017
|
Simport™ Scientific
M5058 |
Tan |
Case of 1500 for $420.00
|
|
|||||
22-045-018
|
Simport™ Scientific
M5059 |
Gray |
Case of 1500 for $420.00
|
|
|||||
22-045-019
|
Simport™ Scientific
M50510 |
Lilac |
Case of 1500 for $432.00
|
|
|||||
22-045-020
|
Simport™ Scientific
M50511 |
Orange |
Case of 1500 for $432.00
|
|
|||||
22-045-021
|
Simport™ Scientific
M50512 |
Aqua |
Case of 1500 for $432.00
|
|
|||||
Description
- 35° Anterior writing area
- One-piece construction with hinge-lock design prevent early separation of base and lid
- Snap latch and hinge-lock design - cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
35° | |
Acetal | |
Tissue Processing/Embedding Cassette |
For in vitro diagnostic (IVD) use only.