Learn More
Cole-Parmer Branson General-purpose ultrasonic cleaning solution, 0.9 L (1 qt) bottle
Supplier: Cole-Parmer 0880111
Safely and effectively remove soils, rust, and oxides from a variety of materials with this cleaning solution. Solutions contain no phosphates and are biodegradable. Dilute with water to the recommended working concentration listed in spec table.