Learn More
OHAUS™ Ultra Flex Support Kit
OHAUS™ Ultra Flex Support Kit includes all the necessary equipment to help when working with hotplate stirrers.
Supplier: OHAUS™ 30400145
Description
- OHAUS offers a range of shaker accessories to meet specific sample requirements
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Tube racks and dimpled mats securely hold tubes in place
Specifications
Ultra Flex Support Kit | |
Clamps, Rods, Sleeves | |
Ultra Flex Support Kit |