Learn More
Revvity Health Sciences Inc Ultima Gold AB, 2x5L
Supplier: Revvity Health Sciences Inc 6013309
Ultima Gold™ AB is the world's first liquid scintillation cocktail specifically designed for alpha/beta discrimination in LSC. Ultima Gold™ AB is based on a unique, high flash-point solvent formulation that provides the slow pulse decay characteristics necessary for effective alpha/beta discrimination. The excellent sample holding capacity of Ultima Gold™ AB for a variety of both aqueous and acidic sample types makes it the cocktail of choice for most alpha/beta LSC applications. Features: Excellent alpha/beta discrimination Excellent for strong mineral acids High quench resistance High sample capacity Biodegradable Very high flash-point of approximately 140'C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.