Learn More
Promega UDP-Glo Glycosyltransferase Assay + UDP-Glucuronic Acid (UDP-GA), 200 assays
The UDP-Glo Glycosyltransferase Assay is a bioluminescent assay for detecting the activity of glycosyltransferases that use UDP-sugars as donor substrates and release UDP as a product.
Supplier: Promega V7061
Description
- Universal Glycosyltransferase Activity Assay
- Easy, “add−incubate−read” protocol
- Detects any glycosyltransferase that uses UDP−sugar as a substrate
- Ideal for low−activity glycosyltransferases
Specifications
Components at different temps | |
200 Assays |