Learn More
Ibidi USA µ-Plate Angiogenesis 96 Well, ibiTreat - Tissue Culture Treated Polymer Coverslip
Supplier: Ibidi USA 89646
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, and µ-Plates which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Plate Angiogenesis 96 Well is a cell culture product for tube formation, angiogenesis assays and direct cell culture in a standard multi-well format. Cells can be grown on or in gel matrices, e.g. Matrigel™ or directly on the ibidi Polymer Coverslip. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.