Learn More
Typenex™ Flexi Band Forms™
Provide perforated inserts for Flexi bands
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
22-355-215
|
Typenex™
FLX001 |
General form | General form | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
22-355-216
|
Typenex™
FLX002 |
Condensed form | Condensed form | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Choose from the general or condensed form, based on the number of stickers required
- With barcoded blood bank ID numbers (BBIDs) for easy scanning into Laboratory Information System (LIS)
- Inserts can be used for handwriting or with patient labels
- Forms have 7-digit BBID in GS1-128 barcode format
- General form provides two patient inserts and 18 barcoded specimen tube stickers
- Condensed form is used for facilities that generate specimen tube stickers as needed; five stickers provided
Specifications
Yes | |
Flexi bands |
Safety and Handling
ShelfLife : 7+ Day(s)