Learn More
Saint Gobain Performance Plastics TYGON TYGN 3/32 x 7/32 50 /PK
Supplier: Saint Gobain Performance Plastics ACF00005
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 1/16 in. Diameter Outer 3/32 in. Maximum Pressure 47 psi at 73 deg.F Thickness 7/32 in. Clear