Learn More
Saint Gobain Performance Plastics Tube Tygon 3/8 x 1/16 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics AAG00027
Encompass_Preferred
Saint-Gobain Tygon Fuel and Lubricant Tubing Formula F-4040-A Diameter Outer 1/2 in. Maximum Pressure 20 psi Thickness 0.0625 in. Diameter Inner 3/8 in. Max. Operating Temperature 74 deg.C Min. Operating Temperature -37 deg.C Overall Length 50 ft. Yellow
Catalog No. 50-206-8929
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product