Learn More
Saint Gobain Performance Plastics Tube Tygon 1/8 x 1/16 50FT-15M/PK
Supplier: Saint Gobain Performance Plastics AAG00007
Saint-Gobain Tygon Fuel and Lubricant Tubing Formula F-4040-A Diameter Outer 1/4 in. Maximum Pressure 40 psi Thickness 0.0625 in. Diameter Inner 0.13 in. Max. Operating Temperature 74 deg.C Min. Operating Temperature -37 deg.C Overall Length 50 ft. Yellow