Learn More
Saint Gobain Performance Plastics Tube Tygon .030 ID 500FT-150M/PK
Supplier:  Saint Gobain Performance Plastics AAD04119
Encompass_Preferred
Saint-Gobain Tygon ND 100-80 Tubing Diameter Inner 0.03 in. Diameter Outer 0.09 in. Thickness 0.03 in. Clear For Use With Medical grade
Catalog No. 50-206-8922
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product