Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 2000 SFR Apron

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Tychem™ 2000 SFR Aprons provide chemical and secondary flame protection in a lightweight garment. Tychem™ 2000 SFR Aprons are intended to be worn over primary flame resistant garments.

  • Tychem™ 2000 SFR represents a new generation of secondary flame-resistant chemical garment technology
  • Unlike traditional secondary flame resistant chemical garments that have been available for years, new Tychem™ 2000 SFR garments were specially designed to meet dual hazard needs of a protective chemical suit with secondary flame resistance
  • This unique combination of performance permits Tychem™ 2000 SFR garments to be worn over primary flame resistant (FR) garments like Nomex™ when chemical splash and flash fire hazards exist
  • The fabric used in Tychem™ 2000 SFR garments utilize a unique technology so that it does not char like traditional secondary flame resistant technologies - it is designed to shrink away from flame without burning
  • Tychem™ 2000 SFR garments also provide an effective barrier against a range of inorganic acids, bases, industrial cleaning chemicals, and particles
  • They're an ideal option for keeping workers safe from chemicals, while not compromising the flame protection of fire-resistance clothing worn beneath them
  • Applications include: refineries, petrochemical plants, laboratories, hazardous maintenance operations
Size Medium
Color Green
Disposable Yes
Closure Type Hook and loop, Waist ties
Seam Type Taped
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Thickness 7.5 mil
Length (English) 43.5 in.
Length (English) Cuff 31.25 in.
Length (Metric) 112 cm
Show More Show Less