Learn More
GFS Chemicals Inc TURBIDITY STD 40 NTU 125ML
Supplier: GFS Chemicals Inc 86231
NTU standard for single detector white light turbidity sensors such as Hach 2100 A, HF Scientific, Orbeco-Hellige, Turner, ICM, , and ESD.