Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Turbidity Standard, 100FNU/FNRU, 60 mL
For use with LaMotte 2020I Turbidity Meter, Infrared Version
Supplier: LaMotte™ 1444
Specifications
LaMotte 2020i Turbidity Meter | |
60 mL | |
2 oz. |
Turbidity Standard, 100 FNU/FBRU, 60 mL | |
100 FNU/FNRU Standard | |
60 mL |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction