Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynalon™ Tuff-Job™ 4-Ply Scrim Reinforced Wipers



Provide superior wipe-dry performance, especially for glass.
Supplier: Dynalon™ 6267150001
This item is not returnable.
View return policy
Description
- Multiple plies for more absorbency
- Polyester scrim webbing for extra strength and durability
- Low lint
- 100% recycled
Specifications
Scrim Reinforced Wipes | |
24.8 cm | |
42.5 cm | |
White | |
Provides superior wipe dry performance, especially for Glass |
9.75 in. | |
16.75 in. | |
Polyester | |
9.75 x 16.75 in. (24.8 x 42.5 cm) | |
4-ply |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title