Learn More
Andwin Scientific TUBES 12X75 FACS CS/1000TUBES
Supplier: Andwin Scientific 60818496/S
The culture polystyrene tubes have round bottoms, volume capacity of 5 mL, and dimension of 12 x 75 mm. The tubes can resist autoclaving temperature up to 122C (252F) and freezing temperature down to -90C (-194F). This product also has an attache