Learn More
Saint Gobain Performance Plastics Tube Tygon 5/32 x 7/32 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAB00009
Encompass_Preferred
Saint-Gobain Tygon S3 Beverage Tubing Formula B-44-3 Overall Length 15.2 m 50 ft. Maximum Pressure 28 psi Diameter Inner 4 mm 0.15 in. Diameter Outer 5.5 mm 0.21 in. Autoclavable Must order minimum quantity of 3.
Catalog No. 50-206-8887
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.