Learn More
Saint Gobain Performance Plastics Tube Tygon 3/4 x 1 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAA00053
Encompass_Preferred
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Non-DEHP Diameter Inner 3/4 in. Diameter Outer 1 in. Maximum Pressure 9.5 MPa
Catalog No. 50-206-8911
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product