Learn More
Saint Gobain Performance Plastics Tube Tygon 3/32 x 5/32 50FT/PK
Supplier: Saint Gobain Performance Plastics AAA00004
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Diameter Inner 0.09 in. 2.4 mm Diameter Outer 0.15 in. 4 mm Autoclavable Autoclavable Certifications/Compliance FDA 21 CFR 175.300 3-A and NSF Standard 51 Hardness 65 (Shore A) Max. Operating Temperature 165 deg.F 79 deg.C Must order minimum quantity of 3.