Learn More
Genesis Bps LLC Tube Sealer Rapid Seal SE 330
Supplier: Genesis Bps LLC 428SE330
Notched Seal™ for easy separation. Fixed sealing head. Designed for heavy duty sealing. RAPID SEAL II™ seals in 0.5 to 1.2 seconds. “Intelligent” RF Board – accepts and adjusts to all size blood tubing. CE and UL compliant. 1 year limited warranty.