Learn More
Andwin Scientific TUBE CULT 13X100 STERILE 1M/CS
Supplier: Andwin Scientific 60818930
The Culture Tubes are constructed with polypropylene. This product is autoclavable, disposable, and sterile. The tubes have a volume capacity of 8 mL, an outer diameter of 13 mm, and length of 100 mm. This is sold in a case of 1000.