Learn More
Saint Gobain Performance Plastics TUB TYGN 1/4 x 1/2 x 1/8 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAB00019
Encompass_Preferred
Saint-Gobain Tygon S3 Beverage Tubing Formula B-44-3 Overall Length 15.2 m 50 ft. Maximum Pressure 60 psi Diameter Inner 6.4 mm 1/4 in. Diameter Outer 12.7 mm 1/2 in. Autoclavable Autoclavable
Catalog No. 50-206-8915
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product