Learn More
Saint Gobain Performance Plastics TUB S-50-HL 1/32 x 3/32 50FT/PK
Supplier:  Saint Gobain Performance Plastics ADF00001
Encompass_Preferred
Saint-Gobain Tygon ND 100-65 Medical Tubing Diameter Inner 1/32 in. 0.8 mm Diameter Outer 3/32 in. 2.4 mm Overall Legth 50 ft. 15.2 m Thickness 1/32 in. Autoclavable Must order minimum quantity of 5
Catalog No. 50-206-8885
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product