Learn More
Saint Gobain Performance Plastics TUB NORPRENE 1/8 x 1/4 50FT/PK
Supplier:  Saint Gobain Performance Plastics AFL00007
Encompass_Preferred
Saint-Gobain Norprene Industrial Tubing Formula A-60-G Diameter Inner 0.13 in. Thickness 0.0625 in. Diameter Outer 3/4 in. Overall Length 50 ft. Black Maximum Pressure 19 psi Must order minimum quantity of 2.
Catalog No. 50-206-8901
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product