Learn More
Thermo Scientific™ TSX ultra-low freezer package with racks, boxes and chart recorder
GREENER_CHOICE
TSX ultra-low freezer package with racks, boxes and factory-installed chart recorder
$30400.00 - $40650.00
Specifications
Type Ultra-Low Freezer, Upright
Temperature Range -50°C to -86°C
Insulation Water-Blown Polyurethane Foam
Interior Stainless Steel
Shelves 3
View More Specs
Alert:
Not available outside North America
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Products 8
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
Catalog Number Mfr. No. Includes Electrical Requirements Type Price Quantity & Availability  
09-313-902
GREENER_CHOICE
Accessories View Documents
Thermo Scientific™
TSX60086DRAKCR
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 208 - 230 V 60 Hz Ultra-Low Freezer, Upright
Each for $36,950.00
Add to Cart
15-500-257
GREENER_CHOICE
View Documents
Thermo Scientific™
TSX40086ARAKCR
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 115 V 60 Hz Ultra-Low Freezer, Upright
Each for $30,400.00
Add to Cart
09-313-881
GREENER_CHOICE
Accessories View Documents
Thermo Scientific™
TSX50086ARAKCR
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 115 V 60 Hz Ultra-Low Freezer, Upright
Each for $34,500.00
Add to Cart
09-313-901
GREENER_CHOICE
Accessories View Documents
Thermo Scientific™
TSX50086DRAKCR
500 box capacity freezer, 10 sliding drawer racks, 250 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 208 - 230 V 60 Hz Ultra-Low Freezer, Upright
Each for $34,500.00
Add to Cart
09-313-882
GREENER_CHOICE
Accessories View Documents
Thermo Scientific™
TSX60086ARAKCR
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 115 V 60 Hz Ultra-Low Freezer, Upright
Each for $36,500.00
Add to Cart
09-313-883
GREENER_CHOICE
Accessories View Documents
Thermo Scientific™
TSX70086ARAKCR
700 box capacity freezer, 14 sliding drawer racks, 350 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 115 V 60 Hz Ultra-Low Freezer, Upright
Each for $40,650.00
Add to Cart
09-313-903
GREENER_CHOICE
Accessories View Documents
Thermo Scientific™
TSX70086DRAKCR
700 box capacity freezer, 14 sliding drawer racks, 350 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 208 - 230 V 60 Hz Ultra-Low Freezer, Upright
Each for $39,750.00
Add to Cart
15-500-258
GREENER_CHOICE
Accessories View Documents
Thermo Scientific™
TSX40086DRAKCR
400 box capacity freezer, 8 sliding drawer racks, 200 2-inch boxes with 100-count cell dividers, and factory-installed chart recorder 208 - 230 V 60 Hz Ultra-Low Freezer, Upright
Each for $30,400.00
Add to Cart
Description
Description
Thermo Scientific™ TSX Ultra-Low temperature freezer package with racks, boxes, and factory-installed chart recorder.
Package includes:
  • One Thermo Scientific TSX Ultra-Low temperature freezer
  • Factory installed chart recorder
  • 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
Warranty and Services
Warranty varies by country, please contact us for details
Specifications
Specifications
Ultra-Low Freezer, Upright
Water-Blown Polyurethane Foam
3
Upright
V-Drive Compressor
-50°C to -86°C
Stainless Steel
EPA SNAP
Alarm, Recorder