Learn More
TrippNT Refrigerator Vented Storage Cabinet
Supplier:  TrippNT 50385
Encompass_Preferred
TrippNT's Refrigerator Vented Storage Cabinet refrigerators or freezers storage with removable trays. The adjustable dividers allows the user create many compartments to fit their own size requirements. Comes with 2 dividers per tray, each tray can hold up to 10 dividers. The distance between each dividers is 1 1/4 Inches. This product comes with 2 removable trays but can hold up to 4 trays. Made from PVC with acrylic dividers this Vented Storage Cabinet is lightweight and easily disinfected, ships fully assembled.
•White Rectangular Design with Open Slats for Ventilation on all Sides
•Adjustable Drawer Dividers
•Constructed from PVC and Blue Acrylic
•Stores Refrigerated Specimens in Removable Drawers
•100% Total Replacement Lifetime Guarantee/Made in USA
Catalog No. 50-856-045