Learn More
Air Science Usa Air Science Safefume Cyanoacrylate Fuming Chamber, CATRI, Three Separate Chambers, 60" / 1500mm Wide, 115V 60Hz
Supplier: Air Science Usa CATRI
Safefume Cyanoacrylate Fuming Chamber (CATRI), 60" Wide, Three Separate Chambers. Safefume cyanoacrylate fuming chambers are designed to safely develop latent fingerprints using ethyl cyanoacrylate (CNA) vapor in a controlled environment for optimum effectiveness and safety where moisture and fuming time are critical factors. The tamper resistant compartment helps maintain the chain of custody. The unique Air Science Multiplex filtration system, together with professional design and unique construction features offer personnel protection during use. The ductless filtration system requires no connection to an outside exhaust system. The automatic control system programs the fuming cycle. A versatile system of hanging rods and shelves allows proper positioning of items of evidence in the chamber. The professionally designed automatic fuming chamber eliminates reliance on do-it-yourself systems that lack user-safety allowances and microprocessor controls needed for repeatable results.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.