Learn More
Wesco Industrial Products Gator Grip Drum Lift for Triple Truck
SDP
Supplier:  Wesco Industrial Products 261161
Encompass_Preferred
Gator Grip Drum Lifter Attachment Handles most 30, 55 and 85 gal. steel, poly and fiber drums with a 3/16" chime.
Catalog No. NC1479359