Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Triple Beam Balance with Weight Set
SCI_ED

Triple beam balance with weighing platform and included weight set.

Supplier:  Walter Products™ B200WO

Catalog No. S139030


Add to cart

Description

Description

  • Rugged die-cast construction
  • Dual magnetic damping system for slow oscillation
  • Included weight set (2 x 1000 g/ 1 x 500 g)
  • Notched and tiered beam with 0.1 g front beam calibration sensitivity
  • Middle beam: 500 g x 100 g
  • Rear beam: 100 g x 10 g
Specifications

Specifications

5.9 in.
15 cm
Balance and Weight Set
2610 g
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit