Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimebutine, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco T615910G
Specifications
39133-31-8 | |
MFCD00133873 | |
387.47 | |
Analytical Standard | |
Analytical Standard |
C22H29 NO5 | |
3,4,5-Trimethoxybenzoic acid 2-(dimethylamino)-2-phenylbutyl ester | |
Neat | |
10 g |
Safety and Handling
Recommended Storage : 2°C to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title