Learn More
Buehler TriDent, PSA, 8 in Polishing Cloth 10/pk
Supplier: Buehler 407518
This item has a minimum qty of 4 per supplier requirements.
TriDent is a soft, synthetic, woven, no nap cloth. It is used to maximize flatness and phase retention on soft to medium-hard materials. Typical applications include but are not limited to ferrous and non-ferrous metals, microelectronics, polymer matrix composites and coatings. 10/pk