Learn More
General Data Healthcare™ TEC™-II Tissue Embedding Center: Cold Plate Console
For use with Triangle Biomedical Services TEC-II Tissue Embedding Center (cat. no. 22046850)
Supplier: General Data Healthcare™ TEC2120CP
Description
- Large, nonstick surface for cooling paraffin embedded blocks
- Temperature can be controlled from 0 to -15°C; All heated surfaces are independently controlled and range in temperature from 50 to 75°C
- Features programmable operating times including automatic start
- 330 x 600 x 405mm (13 x 24 x 16 in.)
Specifications
Cold Plate Console | |
100/120 VAC | |
50/60 Hz |