Learn More
Spectrum™ Trial Size Kits for Biotech-Grade CE Dialysis Membrane Tubing (1 meter), MWCO: 20 kd
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
$230.00 - $273.00
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
08-700-124
|
Spectrum™
131336T |
6.4 mm |
Each for $230.00
|
|
|||||
08-700-126
|
Spectrum™
131348T |
20 mm |
Each for $273.00
|
|
|||||
08-700-125
|
Spectrum™
131345T |
15 mm | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
Kits are great for comparing membrane MWCO performance to optimize separation or purification. Provides a 1m length of Biotech-Grade CE membrane tubing, a pair of universal tubing closures and 5 picks for opening membrane tubing. Tubing is wet packaged in 0.05% sodium azide preservative that easily rinses out with water.
- Rigidly controlled porosity for better selectivity and separations than Standard Grade RC
- Ultrapure, no heavy metal or sulfiide impurities
- Low protein binding
- No glycerin removal, cleaning or pre-treatment required
Specifications
1 m |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.