Learn More
Electron Microscopy Sciences Tri-Pour Beaker Cap for 800 ml beaker
Supplier: Electron Microscopy Sciences 60977
The caps are available for all Tri-Pour beakers. The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
Use: 800 ml