Learn More
Ace Glass, Inc. Trap, body, vacuum, 45/50, 51mm outer body OD, 250mm outer body length below joint
Supplier: Ace Glass, Inc. 875321
Trap, body, vacuum, 45/50, 51mm outer body OD, 250mm outer body length below joint