Learn More
Lab-Aids™ Tracking the Spread of Infectious Diseases
SCI_ED
This activity simulates the transmission of a contagious disease and the steps epidemiologists take to try to determine its source and limit its spread.
Supplier:  Lab-Aids™ 906
Catalog No. S20956
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Teacher's guide; 28 student worksheets and guides; 24 100mL plastic cups; 15mL drop control bottle of 2% sodium carbonate solution; 30mL drop control bottle of "Infection Test Solution"; Transparency: Class Infection History; SDS packet
Description
Description
Contagious diseases can spread quickly through a population. If an infected individual mingles with healthy individuals, random contact can cause many more to become infected. This activity simulates the transmission of a contagious disease and the steps epidemiologists take to try to determine its source and limit its spread. Two scenarios are provided, one for an infection spreading among humans and another among livestock. If desired, the kit materials can also be used to simulate different 'trackable' spreading event scenarios such as an invasive species or an outbreak of food poisoning. This activity also introduces the concept of exponential growth. Accommodates dozens of classes, each with 24 students. Add only cups for larger class sizes. Includes Teacher guide, student sheets, assessment questions and SDS for all chemicals.
Specifications
Specifications
Teacher's guide; 28 student worksheets and guides; 24 100mL plastic cups; 15mL drop control bottle of 2% sodium carbonate solution; 30mL drop control bottle of “Infection Test Solution”; Transparency: Class Infection History; SDS packet
Kit: Tracking the Spread of Infectious Diseases
24 Students