Learn More
TraceCERT™ Rubidium Standard for AAS, 1,000 mg/L Rb in Nitric Acid, MilliporeSigma™ Supelco™
Supplier:  MilliporeSigma Supelco 55727100ML
Catalog No. 11-101-6804
Add to Cart
Description
Description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label.
Specifications
Specifications
7697-37-2
1000 mg/L Rb in nitric acid
100 mL
Certified Reference Material
1000 mg/L Rb in 2% nitric acid, prepared with high purity RbNO3, HNO3 and water
TraceCERT™ Analytical Standard
TraceCERT
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC